6-Chloropyridazine-3-carboxylic acid

Supplier: Matrix Scientific

Synonyms: 3-Carboxy-6-chloropyridazine, 3-Carboxy-6-chloro-1,2-diazine

027325-1G 027325-5G
101823-950EA 29.23 USD
101823-950 101823-952
6-Chloropyridazine-3-carboxylic acid
6-Chloropyridazine-3-carboxylic acid

Hazard: Irritant

Formula: C₅H₃ClN₂O₂
MW: 158.54 g/mol
Melting Pt: 144 °C
MDL Number: MFCD00160464
CAS Number: 5096-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR