3-Bromo-1H-1,2,4-triazole ≥95%

Supplier: Matrix Scientific

027287-1G 027287-5G 027287-25G
101823-892EA 18.46 USD
101823-892 101823-894 102832-960
3-Bromo-1H-1,2,4-triazole ≥95%
3-Bromo-1H-1,2,4-triazole

Hazard: Irritant/Light Sensitive/Stored under Argon

Formula: C₂H₂BrN₃
Melting Pt: 187…189 °C
MDL Number: MFCD01421960
CAS Number: 7343-33-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR