1H-Pyrazolo[3,4-b]pyridin-3-amine

Supplier: Matrix Scientific

Synonyms: 1H-pyrazolo[3,4-b]pyridin-3-amine, 3-Amino-1H-pyrazolo[3,4-b]pyridine

027286-250MG 027286-1G
101823-886EA 387.69 USD
101823-886 101823-888
1H-Pyrazolo[3,4-b]pyridin-3-amine
1H-Pyrazolo[3,4-b]pyridin-3-amine

Hazard: Irritant

Formula: C₆H₆N₄
MW: 134.14 g/mol
MDL Number: MFCD04113611
CAS Number: 6752-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR