4-Methylthiazole-5-carboxylic acid ≥98%

Supplier: Matrix Scientific

027267-5G 027267-25G
101823-848EA 49.23 USD
101823-848 101823-850
4-Methylthiazole-5-carboxylic acid ≥98%
4-Methylthiazole-5-carboxylic acid

Hazard: Irritant

Formula: C₅H₅NO₂S
MW: 143.17 g/mol
MDL Number: MFCD00626872
CAS Number: 20485-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR