3-Amino-N-phenylbenzamide
Supplier: Matrix Scientific
Synonyms:
N-(3-Amino-N-phenyl)benzamide
025016-500MG
025016-1G
101819-410EA
260.23
USD
101819-410
102832-928
3-Amino-N-phenylbenzamide
3-Amino-N-phenylbenzamide
Hazard: Irritant
Formula:
C₁₃H₁₂N₂O MW: 212.25 g/mol |
MDL Number:
MFCD00017101 CAS Number: 14315-16-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...