5-Fluoronicotinic acid ≥97%

Supplier: Matrix Scientific

024948-5G 024948-25G
101819-266EA 36.92 USD
101819-266 101819-268
5-Fluoronicotinic acid ≥97%
5-Fluoronicotinic acid

Hazard: Irritant

Formula: C₆H₄FNO₂
MDL Number: MFCD01318537
CAS Number: 402-66-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR