1,5-Dimethyl-1H-pyrazole ≥97%

Supplier: Matrix Scientific

024851-1G 024851-5G 024851-25G
101819-076EA 15.38 USD
101819-076 101819-078 101819-080
1,5-Dimethyl-1H-pyrazole ≥97%
1,5-Dimethyl-1H-pyrazole

Hazard: Irritant

Formula: C₅H₈N₂
MW: 96.1300 g/mol
MDL Number: MFCD01646129
CAS Number: 694-31-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR