3-Amino-2-pyridone ≥98%

Supplier: Matrix Scientific

024844-5G 024844-25G
101819-052EA 52.31 USD
101819-052 101819-054
3-Amino-2-pyridone ≥98%
3-Amino-2-pyridone

Hazard: Irritant

Formula: C₅H₆N₂O
MDL Number: MFCD03411556
CAS Number: 33630-99-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR