2,6-Dihydroxy-4-(trifluoromethyl)nicotinonitrile ≥97%

Supplier: Matrix Scientific

024835-1G 024835-5G
101819-032EA 18.46 USD
101819-032 101819-034
2,6-Dihydroxy-4-(trifluoromethyl)nicotinonitrile ≥97%
2,6-Dihydroxy-4-(trifluoromethyl)nicotinonitrile

Hazard: Irritant

Formula: C₇H₃F₃N₂O₂
MDL Number: MFCD06200911
CAS Number: 3335-46-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR