(2-Fluoro-5-nitrophenyl)acetic acid ≥98%

Supplier: Matrix Scientific

024824-5G 024824-25G
101819-000EA 43.08 USD
101819-000 101819-002
(2-Fluoro-5-nitrophenyl)acetic acid ≥98%
(2-Fluoro-5-nitrophenyl)acetic acid

Hazard: Irritant

Formula: C₈H₆FNO₄
MDL Number: MFCD04091773
CAS Number: 195609-18-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR