2-Bromo-6-fluorophenol ≥98%
Supplier: Matrix Scientific
024818-5G
024818-25G
101818-984EA
284.62
USD
101818-984
101818-986
2-Bromo-6-fluorophenol ≥98%
2-Bromo-6-fluorophenol
Hazard: Irritant
Formula:
C₆H₄BrFO MW: 191.0000 g/mol |
MDL Number:
MFCD03788533 CAS Number: 2040-89-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...