4-Bromothiophene-2-acetic acid ≥98%

Supplier: Matrix Scientific

Synonyms: (4-Bromothien-2-yl)acetic acid, 2-(4-Bromothiophen-2-yl)acetic acid, 4-Bromo-2-(carboxymethyl)thiophene

024817-1G 024817-5G
101818-980EA 150.77 USD
101818-980 101818-982
4-Bromothiophene-2-acetic acid ≥98%
4-Bromothiophene-2-acetic acid

Hazard: Irritant

Formula: C₆H₅BrO₂S
MDL Number: MFCD09839310
CAS Number: 161942-89-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR