3-Amino-2-fluorobenzonitrile ≥96%

Supplier: Matrix Scientific

024815-1G 024815-5G
101818-972EA 150.77 USD
101818-972 101818-974
3-Amino-2-fluorobenzonitrile ≥96%
3-Amino-2-fluorobenzonitrile

Hazard: Irritant

Formula: C₇H₅FN₂
MDL Number: MFCD08062434
CAS Number: 873697-68-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR