2-Benzyloxy-6-fluorobenzonitrile ≥98%

Supplier: Matrix Scientific

024813-1G 024813-5G 024813-25G
101818-962EA 56.92 USD
101818-962 101818-964 101818-966
2-Benzyloxy-6-fluorobenzonitrile ≥98%
2-Benzyloxy-6-fluorobenzonitrile

Hazard: Irritant

Formula: C₁₄H₁₀FNO
MDL Number: MFCD00068211
CAS Number: 94088-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR