5-Amino-2-methoxy-4-picoline ≥98%

Supplier: Matrix Scientific

Synonyms: 6-Methoxy-4-methylpyridin-3-amine, 5-Amino-2-methoxy-4-methylpyridine

024808-1G 024808-5G 024808-25G
101818-944EA 18.46 USD
101818-944 101818-946 101818-948
5-Amino-2-methoxy-4-picoline ≥98%
5-Amino-2-methoxy-4-picoline

Hazard: Irritant

Formula: C₇H₁₀N₂O
MW: 138.1700 g/mol
MDL Number: MFCD08277274
CAS Number: 6635-91-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR