3-Amino-6-chloro-2-methylpyridine ≥98%

Supplier: Matrix Scientific

024807-1G 024807-5G
101818-940EA 15.38 USD
101818-940 101818-942
3-Amino-6-chloro-2-methylpyridine ≥98%
3-Amino-6-chloro-2-methylpyridine

Hazard: Irritant

Formula: C₆H₇ClN₂
MDL Number: MFCD03095220
CAS Number: 164666-68-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR