2-Bromo-5-methylbenzotrifluoride ≥98%

Supplier: Matrix Scientific

024804-1G 024804-5G
101818-932EA 41.54 USD
101818-932 101818-934
2-Bromo-5-methylbenzotrifluoride ≥98%
2-Bromo-5-methylbenzotrifluoride

Hazard: Irritant

Formula: C₈H₆BrF₃
MW: 239.04 g/mol
MDL Number: MFCD01631586
CAS Number: 261952-20-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR