Methyl pyrimidine-2-carboxylate ≥97%
Supplier: Matrix Scientific
Synonyms:
Pyrimidine-2-carboxylic acid methyll ester
024768-1G
024768-5G
024768-25G
101818-860EA
376.92
USD
101818-860
101818-862
102832-924
Methyl pyrimidine-2-carboxylate ≥97%
Methyl pyrimidine-2-carboxylate
Hazard: Irritant
Formula:
C₆H₆N₂O₂ MW: 138.1200 g/mol |
MDL Number:
MFCD03426929 CAS Number: 34253-03-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...