3-Methyl-1-(2-nitrophenyl)piperazine ≥95%

Supplier: Matrix Scientific

024763-250MG 024763-1G
101818-846EA 153.85 USD
101818-846 101818-848
3-Methyl-1-(2-nitrophenyl)piperazine ≥95%
3-Methyl-1-(2-nitrophenyl)piperazine

Hazard: Irritant

Formula: C₁₁H₁₅N₃O₂
MDL Number: MFCD00448002
CAS Number: 398470-53-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR