2-Pyrimidinecarboxylic acid ≥97%

Supplier: Matrix Scientific

Synonyms: Pyrimidine-2-carboxylic acid, 2-Carboxypyrimidine

024760-1G 024760-5G
101818-836EA 18.46 USD
101818-836 101818-838
2-Pyrimidinecarboxylic acid ≥97%
2-Pyrimidinecarboxylic acid

Hazard: Irritant

Formula: C₅H₄N₂O₂
MW: 124.1 g/mol
MDL Number: MFCD00856161
CAS Number: 31519-62-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR