2-Furane-2-ylpiperazine ≥95%

Supplier: Matrix Scientific

024750-250MG 024750-1G
101818-812EA 392.31 USD
101818-812 101818-814
2-Furane-2-ylpiperazine ≥95%
2-Furane-2-ylpiperazine

Hazard: Irritant

Formula: C₈H₁₂N₂O
MDL Number: MFCD06660430
CAS Number: 111760-37-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR