3-Bromo-4-hydroxyphenylacetic acid ≥98%

Supplier: Matrix Scientific

024747-5G 024747-25G
101818-804EA 23.08 USD
101818-804 101818-806
3-Bromo-4-hydroxyphenylacetic acid ≥98%
3-Bromo-4-hydroxyphenylacetic acid

Hazard: Irritant

Formula: C₈H₇BrO₃
MDL Number: MFCD00016833
CAS Number: 38692-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR