2-Chloro-3-fluoro-5-methylpyridine ≥98%

Supplier: Matrix Scientific

024736-1G 024736-5G
101818-774EA 23.08 USD
101818-774 101818-776
2-Chloro-3-fluoro-5-methylpyridine ≥98%
2-Chloro-3-fluoro-5-methylpyridine

Hazard: Irritant

Formula: C₆H₅ClFN
MW: 145.56 g/mol
MDL Number: MFCD06658238
CAS Number: 34552-15-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR