4-(Difluoromethoxy)chlorobenzene ≥98%

Supplier: Matrix Scientific

Synonyms: 1-Chloro-4-(difluoromethoxy)benzene

024732-5G 024732-25G
101818-760EA 29.23 USD
101818-760 101818-762
4-(Difluoromethoxy)chlorobenzene ≥98%
4-(Difluoromethoxy)chlorobenzene

Hazard: Irritant

Formula: C₇H₅ClF₂O
MW: 178.57 g/mol
MDL Number: MFCD09879256
CAS Number: 81932-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR