5-Bromo-2-fluoro-4-methylpyridine ≥98%

Supplier: Matrix Scientific

024731-5G 024731-25G
101818-756EA 21.54 USD
101818-756 101818-758
5-Bromo-2-fluoro-4-methylpyridine ≥98%
5-Bromo-2-fluoro-4-methylpyridine

Hazard: Irritant

Formula: C₆H₅BrFN
MW: 190.01 g/mol
MDL Number: MFCD09839213
CAS Number: 864830-16-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR