4-Fluoro-3-methoxyphenylacetonitrile ≥98%

Supplier: Matrix Scientific

Synonyms: 2-(4-Fluoro-3-methoxyphenyl)acetonitrile

024728-1G 024728-5G
101818-744EA 43.08 USD
101818-744 101818-746
4-Fluoro-3-methoxyphenylacetonitrile ≥98%
4-Fluoro-3-methoxyphenylacetonitrile

Hazard: Irritant

Formula: C₉H₈FNO
MW: 165.17 g/mol
MDL Number: MFCD09038465
CAS Number: 850565-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR