7-Chloroindole-3-carboxaldehyde ≥98%

Supplier: Matrix Scientific

Synonyms: 7-Chloro-1H-indole-3-carbaldehyde

024709-1G 024709-5G
101818-702EA 141.54 USD
101818-702 101818-704
7-Chloroindole-3-carboxaldehyde ≥98%
7-Chloroindole-3-carboxaldehyde

Hazard: Irritant/Store in Freezer

Formula: C₉H₆ClNO
MW: 179.61 g/mol
MDL Number: MFCD06657153
CAS Number: 1008-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR