6-fluoronicotinonitrile ≥98%

Supplier: Matrix Scientific

Synonyms: 5-Cyano-2-fluoropyridine, 6-Fluoronicotinonitrile

024640-1G 024640-5G
101818-522EA 29.23 USD
101818-522 101818-524
6-fluoronicotinonitrile ≥98%
6-fluoronicotinonitrile

Hazard: Irritant/Irritant-Harmful

Formula: C₆H₃FN₂
MW: 122.102 g/mol
Melting Pt: 51 °C
MDL Number: MFCD09261087
CAS Number: 3939-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR