3,5-Dibromo-2-methoxypyridine ≥98%

Supplier: Matrix Scientific

024630-1G 024630-5G
101818-496EA 21.54 USD
101818-496 101818-498
3,5-Dibromo-2-methoxypyridine ≥98%
3,5-Dibromo-2-methoxypyridine

Hazard: Irritant

Formula: C₆H₅Br₂NO
MW: 266.9200 g/mol
MDL Number: MFCD05663709
CAS Number: 13472-60-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR