3-Amino-6-fluoro-2-methylpyridine ≥98%

Supplier: Matrix Scientific

Synonyms: 6-Fluoro-2-methylpyridin-3-amine

024627-1G 024627-5G
101818-484EA 46.15 USD
101818-484 101818-486
3-Amino-6-fluoro-2-methylpyridine ≥98%
3-Amino-6-fluoro-2-methylpyridine

Hazard: Irritant

Formula: C₆H₇FN₂
MW: 126.13 g/mol
MDL Number: MFCD03094949
CAS Number: 28489-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR