4-Bromothiophene-2-acetonitrile ≥98%

Supplier: Matrix Scientific

024626-5G 024626-25G
101818-480EA 106.15 USD
101818-480 101818-482
4-Bromothiophene-2-acetonitrile ≥98%
4-Bromothiophene-2-acetonitrile

Hazard: Irritant/Irritant-Harmful

Formula: C₆H₄BrNS
MDL Number: MFCD09475422
CAS Number: 160005-43-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR