4-Bromo-2-thiophenecarbonitrile ≥98%

Supplier: Matrix Scientific

Synonyms: 4-Bromothiophene-2-carbonitrile, 4-Bromo-2-cyanothiophene

024617-5G 024617-25G
101818-450EA 86.15 USD
101818-450 101818-452
4-Bromo-2-thiophenecarbonitrile ≥98%
4-Bromo-2-thiophenecarbonitrile

Hazard: Irritant

Formula: C₅H₂BrNS
MW: 188.0500 g/mol
MDL Number: MFCD00114951
CAS Number: 18791-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR