3,5-Dibromo-2-fluoro-6-methylpyridine ≥98%

Supplier: Matrix Scientific

024611-1G 024611-5G
101818-432EA 55.38 USD
101818-432 101818-434
3,5-Dibromo-2-fluoro-6-methylpyridine ≥98%
3,5-Dibromo-2-fluoro-6-methylpyridine

Hazard: Irritant

Formula: C₆H₄NBr₂F
MW: 268.9091 g/mol
MDL Number: MFCD09475419
CAS Number: 632628-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR