3-Amino-5-bromo-2-fluoropyridine ≥98%
Supplier: Matrix Scientific
024610-5G
024610-25G
101818-428EA
67.69
USD
101818-428
101818-430
3-Amino-5-bromo-2-fluoropyridine ≥98%
3-Amino-5-bromo-2-fluoropyridine
Hazard: Irritant
Formula:
C₅H₄BrFN₂ |
MDL Number:
MFCD06659524 CAS Number: 884495-22-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...