5-Bromo-2-hydroxy-3-picoline ≥97%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-3-methylpyridin-2-ol, 5-Bromo-2-hydroxy-3-methylpyridine, 5-Bromo-3-methylpyridin-2(1H)-one

024591-5G 024591-25G
101818-386EA 52.31 USD
101818-386 101818-388
5-Bromo-2-hydroxy-3-picoline ≥97%
5-Bromo-2-hydroxy-3-picoline

Hazard: Irritant

Formula: C₆H₆BrNO
MW: 188.02 g/mol
MDL Number: MFCD03427657
CAS Number: 89488-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR