3-Amino-2-chloro-5-methylpyridine ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-5-methylpyridin-3-amine

024301-5G 024301-25G
101817-828EA 21.54 USD
101817-828 101817-830
3-Amino-2-chloro-5-methylpyridine ≥98%
3-Amino-2-chloro-5-methylpyridine

Hazard: Irritant

Formula: C₆H₇ClN₂
MW: 142.5900 g/mol
MDL Number: MFCD03427656
CAS Number: 34552-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR