Ethyl-3-hydroxybenzoate

Supplier: Matrix Scientific

Synonyms: 3-Hydroxybenzoic acid ethyl ester, Ethyl 3-hydroxybenzoate

024244-25G 024244-100G
101817-706EA 38.46 USD
101817-706 101817-708
Ethyl-3-hydroxybenzoate
Ethyl-3-hydroxybenzoate

Hazard: Irritant

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 187…188 °C (31 mmHg)
Melting Pt: 72…76 °C
MDL Number: MFCD00002296
CAS Number: 7781-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR