4,6-Dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile

Supplier: Matrix Scientific

024219-25G 024219-100G
101817-656EA 49.23 USD
101817-656 101817-658
4,6-Dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile
4,6-Dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile

Hazard: Irritant

Formula: C₈H₈N₂O
MDL Number: MFCD00006269
CAS Number: 769-28-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR