2-Bromo-4-tert-butylphenol ≥97%
Supplier: Matrix Scientific
024167-5G
024167-25G
101817-544EA
32.31
USD
101817-544
101817-546
2-Bromo-4-tert-butylphenol ≥97%
2-Bromo-4-tert-butylphenol
Hazard: Irritant
Formula:
C₁₀H₁₃BrO MW: 229.1100 g/mol |
MDL Number:
MFCD02682891 CAS Number: 2198-66-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...