[4-(Trifluoromethoxy)phenyl]hydrazine
Supplier: Matrix Scientific
027222-5G
027222-25G
101823-762EA
853.85
USD
101823-762
101823-764
[4-(Trifluoromethoxy)phenyl]hydrazine
[4-(Trifluoromethoxy)phenyl]hydrazine
Hazard: Irritant
Formula:
C₇H₇F₃N₂O |
MDL Number:
MFCD02656471 CAS Number: 13957-54-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...