Pyridazine-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

027204-1G 027204-5G
101823-718EA 75.38 USD
101823-718 101823-720
Pyridazine-3-carboxylic acid ≥95%
Pyridazine-3-carboxylic acid

Hazard: Irritant

Formula: C₅H₄N₂O₂
MW: 124.10 g/mol
Melting Pt: ~195°C (dec.)
MDL Number: MFCD01646333
CAS Number: 2164-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR