4-Sulfamoylbenzoic acid

Supplier: Matrix Scientific

Synonyms: 4-Sulphamoylbenzoic acid

027203-25G 027203-100G
101823-714EA 21.54 USD
101823-714 101823-716
4-Sulfamoylbenzoic acid
4-Sulphamoylbenzoic acid

Hazard: Irritant

Formula: C₇H₇NO₄S
MW: 201.2 g/mol
Melting Pt: ca 285°C dec.
MDL Number: MFCD00007938
CAS Number: 138-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR