1,3-Benzothiazole-2-carbaldehyde ≥97%

Supplier: Matrix Scientific

027131-250MG 027131-1G
101823-566EA 252.31 USD
101823-566 101823-568
1,3-Benzothiazole-2-carbaldehyde ≥97%
1,3-Benzothiazole-2-carbaldehyde

Hazard: Irritant

Formula: C₈H₅NOS
MW: 163.2 g/mol
MDL Number: MFCD00526215
CAS Number: 6639-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR