2-Aminobenzo[d]thiazole-6-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Aminobenzothiazole-6-carboxylic acid

027026-1G 027026-5G
101823-346EA 21.54 USD
101823-346 101823-348
2-Aminobenzo[d]thiazole-6-carboxylic acid ≥95%
2-Aminobenzo[d]thiazole-6-carboxylic acid

Hazard: Irritant

Formula: C₈H₆N₂O₂S
MW: 194.21 g/mol
MDL Number: MFCD00054180
CAS Number: 93-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR