4-Fluoro-2-methylbenzenesulfonamide ≥97%

Supplier: Matrix Scientific

026755-1G 026755-5G
101822-808EA 53.85 USD
101822-808 101822-810
4-Fluoro-2-methylbenzenesulfonamide ≥97%
4-Fluoro-2-methylbenzenesulphonamide

Hazard: Irritant

Formula: C₇H₈FNO₂S
MW: 189.2073 g/mol
MDL Number: MFCD02709831
CAS Number: 489-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR