2-Fluoro-3-nitrotoluene ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Fluoro-1-methyl-3-nitrobenzene

026742-25G 026742-100G
101822-796EA 60 USD
101822-796 101822-798
2-Fluoro-3-nitrotoluene ≥98%
2-Fluoro-3-nitrotoluene

Hazard: Irritant/Store in Freezer

Formula: C₇H₆FNO₂
MW: 155.13 g/mol
Boiling Pt: 110-111°C
Melting Pt: 18-19°C
MDL Number: MFCD03412242
CAS Number: 437-86-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR