3-Bromo-5-(trifluoromethyl)pyridin-2-ol ≥98%
Supplier: Matrix Scientific
Synonyms:
3-bromo-5-(trifluoromethyl)pyridin-2-ol, 3-Bromo-2-hydroxy-5-(trifluoromethyl)pyridine, 3-bromo-5-(trifluoromethyl)pyridine-2-ol
026739-1G
026739-5G
101822-782EA
307.69
USD
101822-782
101822-784
3-Bromo-5-(trifluoromethyl)pyridin-2-ol ≥98%
3-Bromo-5-(trifluoromethyl)pyridin-2-ol
Hazard: Irritant
Formula:
C₆H₃BrF₃NO MW: 241.995 g/mol |
MDL Number:
MFCD02691223 CAS Number: 76041-73-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...