2-Amino-3,5-dibromo-4-methylpyridine ≥98%

Supplier: Matrix Scientific

Synonyms: 3,5-Dibromo-4-methylpyridin-2-amine

026738-5G 026738-25G
101822-778EA 21.54 USD
101822-778 101822-780
2-Amino-3,5-dibromo-4-methylpyridine ≥98%
2-Amino-3,5-dibromo-4-methylpyridine

Hazard: Irritant

Formula: C₅H₄Br₂N₂
MDL Number: MFCD01646065
CAS Number: 3430-29-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR