2,3-Dibromoisonicotinic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2,3-Dibromopyridine-4-carboxylic acid

026736-1G 026736-5G
101822-770EA 150.77 USD
101822-770 101822-772
2,3-Dibromoisonicotinic acid ≥95%
2,3-Dibromoisonicotinic acid

Hazard: Irritant

Formula: C₆H₃Br₂NO₂
MW: 280.90 g/mol
MDL Number: MFCD09997708
CAS Number: 1020056-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR