Ethyl-6-(trifluoromethyl)nicotinate ≥98%

Supplier: Matrix Scientific

Synonyms: Ethyl 6-(trifluoromethyl)nicotinate

026735-5G 026735-25G
101822-766EA 84.62 USD
101822-766 101822-768
Ethyl-6-(trifluoromethyl)nicotinate ≥98%
Ethyl-6-(trifluoromethyl)nicotinate

Hazard: Irritant

Formula: C₉H₈F₃NO₂
MW: 219.16 g/mol
MDL Number: MFCD04972865
CAS Number: 597532-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR